N-(2,4-Dinitrophenyl)-6-aminohexanoic acid ≥95.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
6-((2,4-Dinitrophenyl)amino)hexanoic acid, Dnp-ε-Amino-N-Caproic Acid, DNP-X acid, N-Dnp-6-aminohexanoic Acid
D2254-1G
D2254-100MG
TCIAD2254-1GEA
102
EUR
TCIAD2254-1G
TCIAD2254-100MG
N-(2,4-Dinitrophenyl)-6-aminohexanoic acid ≥95.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-6-aminohexanoic acid
Formula:
C₁₂H₁₅N₃O₆ Peso molecolare: 297,27 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00038470 CAS Number: 10466-72-5 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...