Stampa…

2,4-Diamino-6-dimethylamino-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N2,N2-Dimethyl-1,3,5-triazine-2,4,6-triamine, Metformin Related Compound C, N2,N2-Dimethylmelamine

D2236-5G D2236-1G
TCIAD2236-5GEA 221 EUR
TCIAD2236-5G TCIAD2236-1G
2,4-Diamino-6-dimethylamino-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
2,4-Diamino-6-dimethylamino-1,3,5-triazine
Formula: C₅H₁₀N₆
Peso molecolare: 154,18 g/mol
MDL Number: MFCD00191338
CAS Number: 1985-46-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Acetic acid almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR