Stampa…

2,4-Diamino-1,3,5-triazine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3,5-Triazine-2,4-diamine

D2227-1G D2227-5G
TCIAD2227-1GEA 86 EUR
TCIAD2227-1G TCIAD2227-5G
2,4-Diamino-1,3,5-triazine ≥98.0% (by titrimetric analysis)
2,4-Diamino-1,3,5-triazine
Formula: C₃H₅N₅
Peso molecolare: 111,11 g/mol
Punto di fusione: 325°C
MDL Number: MFCD00014598
CAS Number: 504-08-5
EINECS: 207-983-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40ebd1033fe900bc1a3f5d032bbfe60dda444216&vid=40ebd1033fe900bc1a3f5d032bbfe60dda444216&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR