Stampa…

2,11-Dithia[3.3]paracyclophane ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3,10-Dithiatricyclo[10.2.2.2(5,8)]octadeca-5,7,12,14,15,17-hexaene

D2210-100MG D2210-500MG
TCIAD2210-100MGEA 79 EUR
TCIAD2210-100MG TCIAD2210-500MG
2,11-Dithia[3.3]paracyclophane ≥98.0% (by HPLC)
2,11-Dithia[3.3]paracyclophane
Formula: C₁₆H₁₆S₂
Peso molecolare: 272,42 g/mol
MDL Number: MFCD00191410
CAS Number: 28667-63-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 220.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR