meso-2,3-Diphenylsuccinic acid
Fornitore: TCI EUROPE
Synonyms:
meso-2,3-Diphenylbutanedioic Acid
D2209-1G
D2209-5G
TCIAD2209-1GEA
88
EUR
TCIAD2209-1G
TCIAD2209-5G
meso-2,3-Diphenylsuccinic acid
meso-2,3-Diphenylsuccinic acid
Formula:
C₁₆H₁₄O₄ Peso molecolare: 270,28 g/mol |
MDL Number:
MFCD00198082 CAS Number: 1225-13-4 EINECS: 214-953-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...