Stampa…

1,4-Dichloro-2-butanol ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D2194-10G D2194-25G D2194-5G
TCIAD2194-10GEA 272 EUR
TCIAD2194-10G TCIAD2194-25G TCIAD2194-5G
1,4-Dichloro-2-butanol ≥97.0% (by GC)
1,4-Dichloro-2-butanol
Formula: C₄H₈Cl₂O
Peso molecolare: 143,01 g/mol
MDL Number: MFCD00191343
CAS Number: 2419-74-1
EINECS: 219-339-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eca9ced0af08778e4389b47e2f210af4a31cf41b&vid=eca9ced0af08778e4389b47e2f210af4a31cf41b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR