Stampa…

(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate ≥97.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: (-)-2,3:4,6-Di-O-isopropylidene-alpha-L-xylo-2-hexulofuranosonic acid monohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonicacidmonohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate, Dikegulac monohydrate

D2191-25G D2191-5G
TCIAD2191-25GEA 243 EUR
TCIAD2191-25G TCIAD2191-5G
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate ≥97.0% (by titrimetric analysis)
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate
Formula: C₁₂H₁₈O₇·H₂O
Peso molecolare: 274.27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00150517
CAS Number: 68539-16-2
Merck Index: 14,03199

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -19.0 to -22.0 deg (C=2, MeOH)
Water 6.0 to 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR