Stampa…

Ethyl diacetoacetate ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Diacetoacetic Acid Ethyl Ester, Ethyl 2-Acetyl-3-oxobutyrate, Ethyl 2-acetyl-3-oxobutanoate

D2170-25G D2170-5G
TCIAD2170-25GEA 81 EUR
TCIAD2170-25G TCIAD2170-5G
Ethyl diacetoacetate ≥96.0% (by GC)
Ethyl diacetoacetate
Formula: C₈H₁₂O₄
Peso molecolare: 172,18 g/mol
Punto di ebollizione: 103°C
Flash Pt: 85°C
MDL Number: MFCD00009864
CAS Number: 603-69-0
EINECS: 210-053-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR