(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine
D2149-1G
D2149-5G
TCIAD2149-1GEA
47
EUR
TCIAD2149-1G
TCIAD2149-5G
(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥97.0% (by GC)
(3R)-(+)-3-(Dimethylamino)pyrrolidine
Formula:
C₆H₁₄N₂ Peso molecolare: 114,19 g/mol Punto di ebollizione: 166°C Flash Pt: 52°C |
MDL Number:
MFCD00191347 CAS Number: 132958-72-6 UN: 2734 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Specific rotation [a]20/D | +12.0 to +16.0 deg (C=10, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...