Stampa…

ɑ,ɑ'-Dianilino-p-xylene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N,N'-(1,4-Phenylenebis(methylene))dianiline

D2143-1G D2143-5G
TCIAD2143-1GEA 82 EUR
TCIAD2143-1G TCIAD2143-5G
ɑ,ɑ'-Dianilino-p-xylene ≥98.0% (by GC)
alpha,alpha'-Dianilino-p-xylene
Formula: C₂₀H₂₀N₂
Peso molecolare: 288,39 g/mol
MDL Number: MFCD00143578
CAS Number: 13170-62-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR