Stampa…

1,3-Dithiole-2-thione ≥97.0% (by GC)

Fornitore: TCI EUROPE

D2133-5G D2133-1G
TCIAD2133-5GEA 177 EUR
TCIAD2133-5G TCIAD2133-1G
1,3-Dithiole-2-thione ≥97.0% (by GC)
1,3-Dithiole-2-thione
Formula: C₃H₂S₃
Peso molecolare: 134,23 g/mol
MDL Number: MFCD00014536
CAS Number: 930-35-8
EINECS: 213-215-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 97.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR