(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(-)-N,N-Dibutylnorephedrin, (-)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol
D2129-1G
D2129-5G
TCIAD2129-1GEA
51
EUR
TCIAD2129-1G
TCIAD2129-5G
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol ≥98.0% (by GC, titration analysis)
(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol
Formula:
C₁₇H₂₉NO Peso molecolare: 263,43 g/mol |
MDL Number:
MFCD00142695 CAS Number: 114389-70-7 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -22.0 to -26.0 deg (C=2, Hexane) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...