Stampa…

Dimethyl-1,3-dithiole-2-thione-4,5-dicarboxylate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Dimethyl 2-Thioxo-1,3-dithiole-4,5-dicarboxylate, 1,3-Dithiole-2-thione-4,5-dicarboxylic Acid Dimethyl Ester

D2127-10G D2127-25G
TCIAD2127-10GEA 83 EUR
TCIAD2127-10G TCIAD2127-25G
Dimethyl-1,3-dithiole-2-thione-4,5-dicarboxylate ≥98.0% (by GC)
Dimethyl-1,3-dithiole-2-thione-4,5-dicarboxylate
Formula: C₇H₆O₄S₃
Peso molecolare: 250,30 g/mol
MDL Number: MFCD00142532
CAS Number: 7396-41-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 90.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR