Stampa…

Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl Chrysanthemate, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylic Acid Ethyl Ester, Chrysanthemylmonocarboxylic Acid Ethyl Ester, Ethyl Chrysanthemumate

D2112-25G D2112-100G
TCIAD2112-25GEA 32 EUR
TCIAD2112-25G TCIAD2112-100G
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate ≥95.0% (by GC)
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate
Formula: C₁₂H₂₀O₂
Peso molecolare: 196,29 g/mol
Flash Pt: 84°C
MDL Number: MFCD00001304
CAS Number: 97-41-6
EINECS: 202-579-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR