Stampa…

4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3,3'-Dimethylbiphenyl-4,4'-diyl Diisocyanate

D2051-25G D2051-250G
TCIAD2051-25GEA 31 EUR
TCIAD2051-25G TCIAD2051-250G
4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0% (by GC)
4,4'-Diisocyanato-3,3'-dimethylbiphenyl
Formula: C₁₆H₁₂N₂O₂
Peso molecolare: 264,28 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00019908
CAS Number: 91-97-4
EINECS: 202-112-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white pellet
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR