Stampa…

Dess-Martin periodinane ≥95.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

D2045-1G D2045-25G D2045-5G
TCIAD2045-1GEA 43 EUR
TCIAD2045-1G TCIAD2045-25G TCIAD2045-5G
Dess-Martin periodinane ≥95.0% (by titrimetric analysis)
Dess-Martin periodinane
Formula: C₁₃H₁₃IO₈
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1479
ADR: 5.1,III
Merck Index: 14,02933

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR