Stampa…

1,4-Diiodo-2,3,5,6-tetramethylbenzene ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3,6-Diiododurene

D2028-25G D2028-5G
TCIAD2028-25GEA 214 EUR
TCIAD2028-25G TCIAD2028-5G
1,4-Diiodo-2,3,5,6-tetramethylbenzene ≥95.0% (by GC)
1,4-Diiodo-2,3,5,6-tetramethylbenzene
Formula: C₁₀H₁₂I₂
Peso molecolare: 386,01 g/mol
MDL Number: MFCD00142554
CAS Number: 3268-21-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 137.0 to 141.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR