Stampa…

2,5-Diamino-1,4-benzenedithiol dihydrochloride ≥97.0% (by HPLC)

Fornitore: TCI
Attenzione

D2022-25G D2022-5G
TCIAD2022-25GEA 1407 EUR
TCIAD2022-25G TCIAD2022-5G
2,5-Diamino-1,4-benzenedithiol dihydrochloride ≥97.0% (by HPLC)
2,5-Diamino-1,4-benzenedithiol dihydrochloride
Formula: C₆H₈N₂S₂
Punto di fusione: 210°C
MDL Number: MFCD00142835
CAS Number: 75464-52-7

Visualizza i codici

Risultati del test di specificazione

Purity (HPLC) min. 97.0 area%
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR