Stampa…

1,6-Diaminohexane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Hexamethylenediaminetetraacetic Acid

D2019-25G D2019-5G
TCIAD2019-25GEA 586 EUR
TCIAD2019-25G TCIAD2019-5G
1,6-Diaminohexane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
1,6-Diaminohexane-N,N,N',N'-tetraacetic acid
Formula: C₁₄H₂₄N₂O₈
Peso molecolare: 348,35 g/mol
Punto di fusione: 236°C
MDL Number: MFCD00004292
CAS Number: 1633-00-7
EINECS: 216-642-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef0567eaee3d75574403b2ac12665a92747114b&vid=5ef0567eaee3d75574403b2ac12665a92747114b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR