Stampa…

Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulphonate ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2,6-Dinitro-4-(trifluoromethyl)benzenesulphonic acid Sodium Salt

D2011-25G D2011-5G
TCIAD2011-25GEA 436 EUR
TCIAD2011-25G TCIAD2011-5G
Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulphonate ≥95.0% (by titrimetric analysis)
Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulphonate
Formula: C₇H₃F₃N₂O₇S
Peso molecolare: 338,14 g/mol
MDL Number: MFCD00059068
CAS Number: 54495-25-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Ion exchange titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR