Stampa…

Dimethyl(methylthio)sulphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: DMTSF

D1945-1G D1945-5G
TCIAD1945-1GEA 69 EUR
TCIAD1945-1G TCIAD1945-5G
Dimethyl(methylthio)sulphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Dimethyl(methylthio)sulphonium tetrafluoroborate
Formula: C₃H₉BF₄S₂
Peso molecolare: 196,03 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00011806
CAS Number: 5799-67-7
UN: 1759
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR