Stampa…
4,5-Dimethyl-3-hydroxy-2(5H)-furanone ∼14% ∼1.2 mol/L in propylene glycol
Fornitore: TCI
Attenzione
D1851-25ML D1851-5ML
TCIAD1851-25MLEA 145 EUR
TCIAD1851-25ML TCIAD1851-5ML
4,5-Dimethyl-3-hydroxy-2(5H)-furanone ∼14% ∼1.2 mol/L in propylene glycol
4,5-Dimethyl-3-hydroxy-2(5H)-furanone
Formula: C₆H₈O₃
Flash Pt: 104°C
MDL Number: MFCD00059957
CAS Number: 28664-35-9
EINECS: 249-136-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=973c2307ff403e430187698275369d36bc00706f&vid=89d0faea43e5c7ef2a0ec5a3f10fe592642b029e&prid=web&clid=SAPDAM
Visualizza i codici
Concentration (Neutralization titration) 13.0 to 15.0 w/w%