Ethylene glycolbis(3-aminopropyl)ether ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1,2-Bis(3-aminopropoxy)ethane
D1840-25ML
D1840-5ML
D1840-500ML
TCIAD1840-25MLEA
22
EUR
TCIAD1840-25ML
TCIAD1840-5ML
TCIAD1840-500ML
Ethylene glycolbis(3-aminopropyl)ether ≥98.0% (by GC, titration analysis)
Ethylene glycolbis(3-aminopropyl)ether
Formula:
C₈H₂₀N₂O₂ Peso molecolare: 176,26 g/mol Punto di ebollizione: 96°C |
MDL Number:
MFCD00059938 CAS Number: 2997-01-5 EINECS: 221-067-4 UN: 3267 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...