Stampa…

3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

D1817-25G D1817-5G
TCIAD1817-25GEA 144 EUR
TCIAD1817-25G TCIAD1817-5G
3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate ≥98.0% (by HPLC, titration analysis)
3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate
Formula: C₁₀H₁₃NO₄
MDL Number: MFCD00004186
CAS Number: 555-30-6
EINECS: 209-089-2
Merck Index: 14,06055

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -12.5 to -14.5 deg (C=1, H2O)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR