1-(2-Dimethylaminoethyl)-4-methylpiperazine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
D1766-25ML
D1766-500ML
TCIAD1766-25MLEA
16
EUR
TCIAD1766-25ML
TCIAD1766-500ML
1-(2-Dimethylaminoethyl)-4-methylpiperazine ≥98.0% (by GC, titration analysis)
1-(2-Dimethylaminoethyl)-4-methylpiperazine
Formula:
C₉H₂₁N₃ Peso molecolare: 171,29 g/mol Punto di ebollizione: 96°C Flash Pt: 100°C |
MDL Number:
MFCD00059773 CAS Number: 104-19-8 EINECS: 203-183-7 UN: 2922 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...