3-Dimethylamino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Dimethylaminoneopentanol
D1727-500ML
D1727-25ML
TCIAD1727-500MLEA
591
EUR
TCIAD1727-500ML
TCIAD1727-25ML
3-Dimethylamino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Dimethylamino-2,2-dimethyl-1-propanol
Formula:
C₇H₁₇NO Peso molecolare: 131,22 g/mol Punto di ebollizione: 69°C |
MDL Number:
MFCD00059879 CAS Number: 19059-68-8 EINECS: 242-791-7 UN: 2924 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...