(S)-(-)-N,N-Dimethyl-1-phenylethylamine ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₅N Punto di ebollizione: 71°C Flash Pt: 64°C |
MDL Number:
MFCD00056236 CAS Number: 17279-31-1 EINECS: 241-310-8 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -68.0 to -72.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...