Stampa…

2-Diethylaminopyridine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: N-(2-Pyridyl)diethylamine

D1681-25G D1681-5G
TCIAD1681-25GEA 259 EUR
TCIAD1681-25G TCIAD1681-5G
2-Diethylaminopyridine ≥97.0% (by titrimetric analysis)
2-Diethylaminopyridine
Formula: C₉H₁₄N₂
Peso molecolare: 150,23 g/mol
Punto di ebollizione: 118°C
MDL Number: MFCD00059777
CAS Number: 36075-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68034a5bfb04764d40be0be04c86787e9d03ed8d&vid=68034a5bfb04764d40be0be04c86787e9d03ed8d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR