Di(N-succinimidyl) carbonate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
D1662-25G
D1662-5G
TCIAD1662-25GEA
146
EUR
TCIAD1662-25G
TCIAD1662-5G
Di(N-succinimidyl) carbonate ≥98.0% (by titrimetric analysis)
Di(N-succinimidyl) carbonate
Formula:
C₉H₈N₂O₇ Peso molecolare: 256,17 g/mol Punto di fusione: 215°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009767 CAS Number: 74124-79-1 EINECS: 277-730-3 |
Risultati del test di specificazione
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (Butylamine Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...