4,4'-Diaminooctafluorobiphenyl ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,2',3,3',5,5',6,6'-Octafluoro-[1,1'-biphenyl]-4,4'-diamine, Octafluorobenzidine
D1632-5G
D1632-1G
TCIAD1632-5GEA
154
EUR
TCIAD1632-5G
TCIAD1632-1G
4,4'-Diaminooctafluorobiphenyl ≥97.0% (by GC)
4,4'-Diaminooctafluorobiphenyl
Formula:
C₁₂H₄F₈N₂ Peso molecolare: 328,17 g/mol |
MDL Number:
MFCD00007646 CAS Number: 1038-66-0 EINECS: 213-861-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 175.0 to 182.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...