Stampa…

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: ((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene) bis(4-methylbenzenesulphonate), (4S,5S)-(-)-2,3-O-Isopropylidene-1,4-di-O-tosyl-L-threitol, (4S,5S)-(+)-O-Isopropylidene-2,3-dihydroxy-1,4-bis(p-tosyl)butane

D1623-1G D1623-5G
TCIAD1623-1GEA 38 EUR
TCIAD1623-1G TCIAD1623-5G
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol ≥98.0% (by HPLC)
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol
Formula: C₂₁H₂₆O₈S₂
Peso molecolare: 470,55 g/mol
MDL Number: MFCD00003212
CAS Number: 37002-45-2
EINECS: 253-306-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR