Stampa…

4-Dimethylaminobenzylamine dihydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-(Aminomethyl)-N,N-dimethylaniline dihydrochloride

D1620-25G D1620-5G
TCIAD1620-25GEA 489 EUR
TCIAD1620-25G TCIAD1620-5G
4-Dimethylaminobenzylamine dihydrochloride ≥98.0% (by titrimetric analysis)
4-Dimethylaminobenzylamine dihydrochloride
Formula: C₉H₁₄N₂
MDL Number: MFCD00012862
CAS Number: 34403-52-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR