Dichloromethylenedimethyliminium chloride ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₃H₆Cl₃N |
MDL Number:
MFCD00011870 CAS Number: 33842-02-3 EINECS: 251-695-4 UN: 3261 ADR: 8,III Merck Index: 14,09977 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...