2,5-Dimethyl-4-hydroxy-3(2H)-furanone ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, Furaneol, 4-Hydroxy-2,5-dimethylfuran-3(2H)-one
D1569-25G
D1569-5G
TCIAD1569-25GEA
81
EUR
TCIAD1569-25G
TCIAD1569-5G
2,5-Dimethyl-4-hydroxy-3(2H)-furanone ≥98.0% (by HPLC)
2,5-Dimethyl-4-hydroxy-3(2H)-furanone
Formula:
C₆H₈O₃ Peso molecolare: 128,13 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00010706 CAS Number: 3658-77-3 EINECS: 222-908-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...