2,3-Dihydro-2-methylbenzofuran ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Methyl-2,3-dihydrobenzofuran
D1537-10G
D1537-25G
TCIAD1537-10GEA
108
EUR
TCIAD1537-10G
TCIAD1537-25G
2,3-Dihydro-2-methylbenzofuran ≥98.0% (by GC)
2,3-Dihydro-2-methylbenzofuran
Formula:
C₉H₁₀O Peso molecolare: 134,18 g/mol Punto di ebollizione: 198°C Flash Pt: 63°C |
MDL Number:
MFCD00023003 CAS Number: 1746-11-8 EINECS: 217-124-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...