Dansyl-L-valine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Dns-Val-OH, 5-Dimethylaminonaphthalene-1-sulphonyl-L-valine
D1505-1G
D1505-100MG
TCIAD1505-1GEA
234
EUR
TCIAD1505-1G
TCIAD1505-100MG
Dansyl-L-valine ≥98.0% (by titrimetric analysis)
Dansyl-L-valine
Formula:
C₁₇H₂₂N₂O₄S Peso molecolare: 350,43 g/mol |
MDL Number:
MFCD00069524 CAS Number: 1098-50-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 137.0 to 140.0 deg-C |
Specific rotation [a]20/D | +18.0 to +21.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...