Stampa…

Dansyl-L-phenylalanine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Dns-Phe-OH, 5-Dimethylaminonaphthalene-1-sulphonyl-L-phenylalanine

D1500-1G D1500-5G D1500-100MG
TCIAD1500-1GEA 87 EUR
TCIAD1500-1G TCIAD1500-5G TCIAD1500-100MG
Dansyl-L-phenylalanine ≥98.0% (by titrimetric analysis)
Dansyl-L-phenylalanine
Formula: C₂₁H₂₂N₂O₄S
Peso molecolare: 398,48 g/mol
MDL Number: MFCD00037730
CAS Number: 1104-36-5
EINECS: 214-165-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR