Dodecylamine hydrochloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Dodecan-1-amine hydrochloride, Laurylamine Hydrochloride
Formula:
C₁₂H₂₇N Peso molecolare: 221.81 g/mol |
MDL Number:
MFCD00042017 CAS Number: 929-73-7 EINECS: 213-205-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...