Stampa…

Sodium trifluoroacetate ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Sodium 2,2,2-trifluoroacetate

A14613.14 A14613.22 A14613.36
A14613.14EA 37 EUR
A14613.14 A14613.22 A14613.36
Sodium trifluoroacetate ≥98%
Sodium trifluoroacetate

Purity: 98% CAS No.: 2923-18-4 Molecular Formula: CF3CO2Na Formula Weight: 136.00 Melting Point: ca 207° dec. Beilstein Reference: 3597949

Formula: C₂F₃NaO₂
Peso molecolare: 136,01 g/mol
Punto di fusione: ∼ 207 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013217
CAS Number: 2923-18-4
EINECS: 220-879-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR