Stampa…

5,5-Diphenylhydantoin sodium salt ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Phenytoin sodium, Sodium 5,5-Diphenylhydantoin, Sodium 2,4-dioxo-5,5-diphenylimidazolidin-1-ide

D1331-25G D1331-500G
TCIAD1331-25GEA 20 EUR
TCIAD1331-25G TCIAD1331-500G
5,5-Diphenylhydantoin sodium salt ≥98.0% (by titrimetric analysis)
5,5-Diphenylhydantoin sodium salt
Formula: C₁₅H₁₂N₂O₂
Peso molecolare: 274,25 g/mol
MDL Number: MFCD00069674
CAS Number: 630-93-3
EINECS: 211-148-2
Merck Index: 14,07322

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR