Stampa…

Decamethonium bromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: Decane-1,10-bis(trimethylammonium Bromide), Decamethonium Bromide, Decamethylenebis(trimethylammonium Bromide)

D1314-25G D1314-5G
TCIAD1314-25GEA 221 EUR
TCIAD1314-25G TCIAD1314-5G
Decamethonium bromide ≥98.0% (by titrimetric analysis)
Decamethonium bromide
Formula: C₁₆H₃₈Br₂N₂
MDL Number: MFCD00011779
CAS Number: 541-22-0
EINECS: 208-772-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,02847
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57868f9b896b2074994eccb1a51fa9bc2b10e3ea&vid=57868f9b896b2074994eccb1a51fa9bc2b10e3ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR