Stampa…

N,N-Dimethylaminomethylferrocene ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Ferrocenylmethyldimethylamine, (Dimethylaminomethyl)ferrocene

D1271-25G D1271-5G
TCIAD1271-25GEA 107 EUR
TCIAD1271-25G TCIAD1271-5G
N,N-Dimethylaminomethylferrocene ≥97.0% (by GC, titration analysis)
N,N-Dimethylaminomethylferrocene
Formula: C₁₃H₁₇FeN
Peso molecolare: 243,13 g/mol
MDL Number: MFCD00001433
CAS Number: 1271-86-9
EINECS: 215-044-8

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown clear liquid to cloudy liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR