1-(3-Dimethylaminophenyl)ethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
1-(3-Dimethylaminophenyl)ethyl Alcohol, 3-Dimethylamino-alpha-methylbenzyl Alcohol
D1269-25G
D1269-5G
TCIAD1269-25GEA
401
EUR
TCIAD1269-25G
TCIAD1269-5G
1-(3-Dimethylaminophenyl)ethanol ≥98.0% (by GC)
1-(3-Dimethylaminophenyl)ethanol
Formula:
C₁₀H₁₅NO Peso molecolare: 165,24 g/mol Punto di ebollizione: 102°C |
MDL Number:
MFCD00059658 CAS Number: 5339-01-5 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...