Diethylene glycol diacetate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Diethyleneglycoldiacetate, Oxybis(2-ethyl Acetate)
Formula:
C₈H₁₄O₅ Peso molecolare: 190.20 g/mol Flash Pt: 135°C |
MDL Number:
MFCD00026211 CAS Number: 628-68-2 EINECS: 211-049-4 |
Risultati del test di specificazione
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...