3,5-Diiodo-L-thyronine ≥96.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
3,5-Diiodo-L-thyronine, (S)-2-Amino-3-(4-(4-hydroxyphenoxy)-3,5-diiodophenyl)propanoic acid
D1119-1G
D1119-100MG
D1119-5G
TCIAD1119-1GEA
114
EUR
TCIAD1119-1G
TCIAD1119-100MG
TCIAD1119-5G
3,5-Diiodo-L-thyronine ≥96.0% (by titrimetric analysis)
3,5-Diiodo-L-thyronine
Formula:
C₁₅H₁₃I₂NO₄ Peso molecolare: 525,08 g/mol |
MDL Number:
MFCD00064987 CAS Number: 1041-01-6 EINECS: 213-867-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Specific rotation [a]20/D | +26.0 to +30.0 deg (C=1, 1mol/L HCl:EtOH=1:2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...