N,N-Dimethylacrylamide ≥99.0% (by GC) stabilizzato
Fornitore: TCI
|
Pericolo
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₅H₉NO Punto di ebollizione: 80°C Flash Pt: 77°C |
MDL Number:
MFCD00008626 CAS Number: 2680-03-7 EINECS: 220-237-5 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Bromination Method) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...