Nɑ,N-ε-Bis(2,4-dinitrophenyl)-L-lysine ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Nɑ,N-ε-Di-Dnp-L-lysine, Dnp-Lys(Dnp)-OH
D1051-1G
D1051-100MG
TCIAD1051-1GEA
153
EUR
TCIAD1051-1G
TCIAD1051-100MG
Nɑ,N-ε-Bis(2,4-dinitrophenyl)-L-lysine ≥97.0% (by HPLC)
Nalpha,N-epsilon-Bis(2,4-dinitrophenyl)-L-lysine
Formula:
C₁₈H₁₈N₆O₁₀ Peso molecolare: 478,37 g/mol |
MDL Number:
MFCD00056105 CAS Number: 1655-49-8 EINECS: 216-737-8 UN: 1325 ADR: 4.1,II |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 174.0 to 178.0 deg-C |
Specific rotation [a]20/D | -30.0 to -37.0 deg (C=0.2, AcOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...