N-(2,4-Dinitrophenyl)-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Dnp-Phe-OH, N-Dnp-L-phenylalanine
Formula:
C₁₅H₁₃N₃O₆ Peso molecolare: 331.28 g/mol |
MDL Number:
MFCD00038099 CAS Number: 1655-54-5 EINECS: 216-742-5 UN: 1325 ADR: 4.1 II |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Specific rotation [a]20/D | -90.0 to -99.0 deg (C=1, AcOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...