Stampa…

Methyl cis-13-docosenoate ≥90.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: cis-13-Docosenoic Acid Methyl Ester, Methyl Erucate, (Z)-Methyl docos-13-enoate, Erucic Acid Methyl Ester

D1017-25ML D1017-500ML
TCIAD1017-25MLEA 48 EUR
TCIAD1017-25ML TCIAD1017-500ML
Methyl cis-13-docosenoate ≥90.0% (by GC)
Methyl cis-13-docosenoate
Formula: C₂₃H₄₄O₂
Peso molecolare: 352,60 g/mol
Punto di ebollizione: 222°C
Flash Pt: 33°C
MDL Number: MFCD00027343
CAS Number: 1120-34-9
EINECS: 214-305-3
UN: 3272
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR