Stampa…

Dodecylbenzene (mixture of branched chain isomers), hard type

Fornitore: TCI EUROPE

Synonyms: Alkylbenzene, Laurylbenzene

D0988-25ML D0988-500ML
TCIAD0988-25MLEA 17 EUR
TCIAD0988-25ML TCIAD0988-500ML
Dodecylbenzene (mixture of branched chain isomers), hard type
Dodecylbenzene (mixture of branched chain isomers)
Formula: C₁₈H₃₀
Peso molecolare: 246,44 g/mol
Flash Pt: 124°C
MDL Number: MFCD00008974
CAS Number: 25265-78-5
EINECS: 246-772-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) multi component mixture
Average alkylchain length 11.0 to 13.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR